Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHCGR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| LHCGR - luteinizing hormone/choriogonadotropin receptor | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000233.3 | Intron | NP_000224.2 | ||||
| XM_005264309.3 | Intron | XP_005264366.1 | ||||
| XM_006712015.3 | Intron | XP_006712078.1 | ||||
| XM_011532828.1 | Intron | XP_011531130.1 | ||||
| XM_011532831.1 | Intron | XP_011531133.1 | ||||
| XM_011532834.2 | Intron | XP_011531136.1 | ||||
| XM_017004089.1 | Intron | XP_016859578.1 | ||||
| XM_017004090.1 | Intron | XP_016859579.1 | ||||
| STON1-GTF2A1L - STON1-GTF2A1L readthrough | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001198593.1 | Intron | NP_001185522.1 | ||||
| NM_001198594.1 | Intron | NP_001185523.1 | ||||
| NM_172311.2 | Intron | NP_758515.1 | ||||

LHCGR
Genomic Map