Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6842 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| MIR6842 - microRNA 6842 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PTK2B - protein tyrosine kinase 2 beta | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004103.4 | Intron | NP_004094.3 | ||||
| NM_173174.2 | Intron | NP_775266.1 | ||||
| NM_173175.2 | Intron | NP_775267.1 | ||||
| NM_173176.2 | Intron | NP_775268.1 | ||||
| XM_005273447.4 | Intron | XP_005273504.1 | ||||
| XM_005273448.4 | Intron | XP_005273505.1 | ||||
| XM_011544441.2 | Intron | XP_011542743.1 | ||||
| XM_011544442.2 | Intron | XP_011542744.1 | ||||
| XM_017013214.1 | Intron | XP_016868703.1 | ||||
| XM_017013215.1 | Intron | XP_016868704.1 | ||||
| XM_017013216.1 | Intron | XP_016868705.1 | ||||

MIR6842
Genomic Map