Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUMA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| NUMA1 - nuclear mitotic apparatus protein 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001286561.1 | Intron | NP_001273490.1 | ||||
| NM_006185.3 | Intron | NP_006176.2 | ||||
| XM_006718564.1 | Intron | XP_006718627.1 | ||||
| XM_011545054.1 | Intron | XP_011543356.1 | ||||
| XM_011545055.1 | Intron | XP_011543357.1 | ||||
| XM_011545056.2 | Intron | XP_011543358.1 | ||||
| XM_011545057.2 | Intron | XP_011543359.1 | ||||
| XM_011545058.1 | Intron | XP_011543360.1 | ||||
| XM_011545059.2 | Intron | XP_011543361.1 | ||||
| XM_011545060.2 | Intron | XP_011543362.1 | ||||
| XM_011545061.2 | Intron | XP_011543363.1 | ||||
| XM_011545062.2 | Intron | XP_011543364.1 | ||||
| XM_011545063.2 | Intron | XP_011543365.1 | ||||
| XM_011545064.1 | Intron | XP_011543366.1 | ||||
| XM_011545065.1 | Intron | XP_011543367.1 | ||||
| XM_011545066.1 | Intron | XP_011543368.1 | ||||
| XM_017017830.1 | Intron | XP_016873319.1 | ||||
| XM_017017831.1 | Intron | XP_016873320.1 | ||||

NUMA1
Genomic Map