Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| RGPD1 - RANBP2-like and GRIP domain containing 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001024457.3 | Intron | NP_001019628.3 | ||||
| XM_011532845.2 | Intron | XP_011531147.1 | ||||
| XM_011532849.2 | Intron | XP_011531151.1 | ||||
| XM_011532850.1 | Intron | XP_011531152.1 | ||||
| XM_011532851.2 | Intron | XP_011531153.1 | ||||
| XM_011532852.2 | Intron | XP_011531154.1 | ||||
| XM_017004103.1 | Intron | XP_016859592.1 | ||||
| XM_017004104.1 | Intron | XP_016859593.1 | ||||
| XM_017004105.1 | Intron | XP_016859594.1 | ||||
| XM_017004106.1 | Intron | XP_016859595.1 | ||||
| XM_017004107.1 | Intron | XP_016859596.1 | ||||

RGPD1
Genomic Map