Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MIR153-2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MIR153-2 - microRNA 153-2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PTPRN2 - protein tyrosine phosphatase, receptor type N2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001308267.1 | 2656 | Silent Mutation | GTA,GTG | V,V 803 | NP_001295196.1 | |
| NM_001308268.1 | 2656 | Silent Mutation | GTA,GTG | V,V 864 | NP_001295197.1 | |
| NM_002847.4 | 2656 | Silent Mutation | GTA,GTG | V,V 841 | NP_002838.2 | |
| NM_130842.3 | 2656 | Silent Mutation | GTA,GTG | V,V 824 | NP_570857.2 | |
| NM_130843.3 | 2656 | Silent Mutation | GTA,GTG | V,V 812 | NP_570858.2 | |
| XM_011516446.1 | 2656 | Intron | XP_011514748.1 | |||
| XM_011516447.2 | 2656 | Intron | XP_011514749.1 | |||
| XM_011516448.2 | 2656 | Intron | XP_011514750.1 | |||
| XM_011516449.2 | 2656 | Intron | XP_011514751.1 | |||
| XM_017012475.1 | 2656 | Silent Mutation | GTA,GTG | V,V 775 | XP_016867964.1 | |
| XM_017012476.1 | 2656 | Intron | XP_016867965.1 | |||

MIR153-2
Genomic Map