Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| GLRA2 - glycine receptor alpha 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001118885.1 | Intron | NP_001112357.1 | ||||
| NM_001118886.1 | Intron | NP_001112358.1 | ||||
| NM_001171942.1 | Intron | NP_001165413.1 | ||||
| NM_002063.3 | Intron | NP_002054.1 | ||||
| XM_006724487.3 | Intron | XP_006724550.1 | ||||
| XM_011545495.2 | Intron | XP_011543797.1 | ||||
| XM_011545496.2 | Intron | XP_011543798.1 | ||||
| XM_017029427.1 | Intron | XP_016884916.1 | ||||
| XM_017029428.1 | Intron | XP_016884917.1 | ||||
| XM_017029429.1 | Intron | XP_016884918.1 | ||||

GLRA2
Genomic Map