Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
NLRP14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| NLRP14 - NLR family pyrin domain containing 14 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_176822.3 | Intron | NP_789792.1 | ||||
| XM_011520044.1 | Intron | XP_011518346.1 | ||||
| ZNF214 - zinc finger protein 214 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_013249.2 | Intron | NP_037381.2 | ||||
| XM_005253128.3 | Intron | XP_005253185.1 | ||||
| XM_005253129.3 | Intron | XP_005253186.1 | ||||
| XM_006718308.3 | Intron | XP_006718371.1 | ||||
| XM_006718309.3 | Intron | XP_006718372.1 | ||||
| XM_011520355.2 | Intron | XP_011518657.1 | ||||
| XM_011520357.2 | Intron | XP_011518659.1 | ||||
| XM_017018269.1 | Intron | XP_016873758.1 | ||||

NLRP14
Genomic Map