Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ANO3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ANO3 - anoctamin 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001313726.1 | Intron | NP_001300655.1 | ||||
| NM_001313727.1 | Intron | NP_001300656.1 | ||||
| NM_031418.3 | Intron | NP_113606.2 | ||||
| XM_011520282.2 | Intron | XP_011518584.1 | ||||
| XM_017018118.1 | Intron | XP_016873607.1 | ||||
| XM_017018119.1 | Intron | XP_016873608.1 | ||||
| MUC15 - mucin 15, cell surface associated | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001135091.1 | Intron | NP_001128563.1 | ||||
| NM_001135092.1 | Intron | NP_001128564.1 | ||||
| NM_145650.3 | Intron | NP_663625.2 | ||||
| XM_017017235.1 | Intron | XP_016872724.1 | ||||
| XM_017017236.1 | Intron | XP_016872725.1 | ||||

ANO3
Genomic Map