Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GABPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GABPA - GA binding protein transcription factor alpha subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001197297.1 | Intron | NP_001184226.1 | ||||
| NM_002040.3 | Intron | NP_002031.2 | ||||
| XM_005260938.4 | Intron | XP_005260995.1 | ||||
| XM_011529520.2 | Intron | XP_011527822.1 | ||||
| XM_011529521.2 | Intron | XP_011527823.1 | ||||
| XM_017028313.1 | Intron | XP_016883802.1 | ||||

GABPA
Genomic Map