Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ATP5J PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ATP5J - ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001003696.1 | 488 | Missense Mutation | ATG,ATT | M,I 47 | NP_001003696.1 | |
| NM_001003697.1 | 488 | Missense Mutation | ATG,ATT | M,I 47 | NP_001003697.1 | |
| NM_001003701.1 | 488 | Missense Mutation | ATG,ATT | M,I 55 | NP_001003701.1 | |
| NM_001003703.1 | 488 | Missense Mutation | ATG,ATT | M,I 47 | NP_001003703.1 | |
| NM_001320266.1 | 488 | Missense Mutation | ATG,ATT | M,I 47 | NP_001307195.1 | |
| NM_001320267.1 | 488 | Missense Mutation | ATG,ATT | M,I 47 | NP_001307196.1 | |
| NM_001685.4 | 488 | Missense Mutation | ATG,ATT | M,I 47 | NP_001676.2 | |
| GABPA - GA binding protein transcription factor alpha subunit | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

ATP5J
Genomic Map