Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GABRA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GABRA3 - gamma-aminobutyric acid type A receptor alpha3 subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000808.3 | Intron | NP_000799.1 | ||||
| XM_006724811.2 | Intron | XP_006724874.1 | ||||

GABRA3
Genomic Map