Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
RANBP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| RANBP2 - RAN binding protein 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_006267.4 | Intron | NP_006258.3 | ||||
| XM_005264002.2 | Intron | XP_005264059.1 | ||||
| XM_005264003.2 | Intron | XP_005264060.1 | ||||
| XM_005264004.2 | Intron | XP_005264061.1 | ||||
| XM_005264005.4 | Intron | XP_005264062.1 | ||||
| XM_005264007.2 | Intron | XP_005264064.1 | ||||
| XM_011511575.2 | Intron | XP_011509877.1 | ||||
| XM_011511576.2 | Intron | XP_011509878.1 | ||||
| XM_011511578.2 | Intron | XP_011509880.1 | ||||
| XM_017004623.1 | Intron | XP_016860112.1 | ||||
| XM_017004624.1 | Intron | XP_016860113.1 | ||||
| XM_017004625.1 | Intron | XP_016860114.1 | ||||
| SH3RF3 - SH3 domain containing ring finger 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001099289.2 | Intron | NP_001092759.1 | ||||
| XM_011511109.2 | Intron | XP_011509411.1 | ||||
| XM_011511110.2 | Intron | XP_011509412.1 | ||||

RANBP2
Genomic Map