Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
DPP10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| DPP10 - dipeptidyl peptidase like 10 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001004360.3 | Intron | NP_001004360.2 | ||||
| NM_001178034.1 | Intron | NP_001171505.1 | ||||
| NM_001178036.1 | Intron | NP_001171507.1 | ||||
| NM_001178037.1 | Intron | NP_001171508.1 | ||||
| NM_001321905.1 | Intron | NP_001308834.1 | ||||
| NM_001321906.1 | Intron | NP_001308835.1 | ||||
| NM_001321907.1 | Intron | NP_001308836.1 | ||||
| NM_001321908.1 | Intron | NP_001308837.1 | ||||
| NM_001321909.1 | Intron | NP_001308838.1 | ||||
| NM_001321910.1 | Intron | NP_001308839.1 | ||||
| NM_001321911.1 | Intron | NP_001308840.1 | ||||
| NM_001321912.1 | Intron | NP_001308841.1 | ||||
| NM_001321913.1 | Intron | NP_001308842.1 | ||||
| NM_001321914.1 | Intron | NP_001308843.1 | ||||
| NM_020868.4 | Intron | NP_065919.2 | ||||
| XM_017004565.1 | Intron | XP_016860054.1 | ||||
| XM_017004566.1 | Intron | XP_016860055.1 | ||||

DPP10
Genomic Map