Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
CNTN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| CNTN4 - contactin 4 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001206955.1 | Intron | NP_001193884.1 | ||||
| NM_001206956.1 | Intron | NP_001193885.1 | ||||
| NM_175607.2 | Intron | NP_783200.1 | ||||
| NM_175613.2 | Intron | NP_783302.1 | ||||
| XM_006713004.3 | Intron | XP_006713067.1 | ||||
| XM_011533425.2 | Intron | XP_011531727.1 | ||||
| XM_011533426.2 | Intron | XP_011531728.1 | ||||
| XM_011533427.2 | Intron | XP_011531729.1 | ||||
| XM_011533428.2 | Intron | XP_011531730.1 | ||||
| XM_011533429.2 | Intron | XP_011531731.1 | ||||
| XM_011533430.2 | Intron | XP_011531732.1 | ||||
| XM_011533431.2 | Intron | XP_011531733.1 | ||||
| XM_017005782.1 | Intron | XP_016861271.1 | ||||
| XM_017005783.1 | Intron | XP_016861272.1 | ||||
| XM_017005784.1 | Intron | XP_016861273.1 | ||||
| XM_017005785.1 | Intron | XP_016861274.1 | ||||
| XM_017005786.1 | Intron | XP_016861275.1 | ||||
| XM_017005787.1 | Intron | XP_016861276.1 | ||||
| XM_017005788.1 | Intron | XP_016861277.1 | ||||
| CNTN4-AS1 - CNTN4 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

CNTN4
Genomic Map