Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
RARB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| RARB - retinoic acid receptor beta | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000965.4 | Intron | NP_000956.2 | ||||
| NM_001290216.2 | Intron | NP_001277145.1 | ||||
| NM_001290217.1 | Intron | NP_001277146.1 | ||||
| NM_001290266.1 | Intron | NP_001277195.1 | ||||
| NM_001290276.1 | Intron | NP_001277205.1 | ||||
| NM_001290277.1 | Intron | NP_001277206.1 | ||||
| NM_001290300.1 | Intron | NP_001277229.1 | ||||
| NM_016152.3 | Intron | NP_057236.1 | ||||

RARB
Genomic Map