Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ROBO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ROBO1 - roundabout guidance receptor 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001145845.1 | Intron | NP_001139317.1 | ||||
| NM_002941.3 | Intron | NP_002932.1 | ||||
| NM_133631.3 | Intron | NP_598334.2 | ||||
| XM_006713277.2 | Intron | XP_006713340.1 | ||||
| XM_011533976.1 | Intron | XP_011532278.1 | ||||
| XM_011533977.2 | Intron | XP_011532279.1 | ||||
| XM_011533978.1 | Intron | XP_011532280.1 | ||||
| XM_011533979.1 | Intron | XP_011532281.1 | ||||
| XM_011533980.1 | Intron | XP_011532282.1 | ||||
| XM_017006982.1 | Intron | XP_016862471.1 | ||||
| XM_017006983.1 | Intron | XP_016862472.1 | ||||
| XM_017006984.1 | Intron | XP_016862473.1 | ||||
| XM_017006985.1 | Intron | XP_016862474.1 | ||||

ROBO1
Genomic Map