Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
DBN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| DBN1 - drebrin 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| GRK6 - G protein-coupled receptor kinase 6 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PRR7 - proline rich 7 (synaptic) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001174101.1 | 1566 | Intron | NP_001167572.1 | |||
| NM_001174102.1 | 1566 | Intron | NP_001167573.1 | |||
| NM_030567.4 | 1566 | Intron | NP_085044.2 | |||
| XM_011534662.1 | 1566 | UTR 5 | XP_011532964.1 | |||
| XM_011534663.2 | 1566 | Intron | XP_011532965.1 | |||
| XM_017009896.1 | 1566 | UTR 5 | XP_016865385.1 | |||
| XM_017009897.1 | 1566 | Intron | XP_016865386.1 | |||
| PRR7-AS1 - PRR7 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

DBN1
Genomic Map