Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GABRA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GABRA1 - gamma-aminobutyric acid type A receptor alpha1 subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000806.5 | 1239 | Missense Mutation | ACA,ATA | T,I 295 | NP_000797.2 | |
| NM_001127643.1 | 1239 | Missense Mutation | ACA,ATA | T,I 295 | NP_001121115.1 | |
| NM_001127644.1 | 1239 | Missense Mutation | ACA,ATA | T,I 295 | NP_001121116.1 | |
| NM_001127645.1 | 1239 | Missense Mutation | ACA,ATA | T,I 295 | NP_001121117.1 | |
| NM_001127648.1 | 1239 | Missense Mutation | ACA,ATA | T,I 295 | NP_001121120.1 | |

GABRA1
Genomic Map