Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MICAL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MICAL1 - microtubule associated monooxygenase, calponin and LIM domain containing 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001159291.1 | 1520 | Intron | NP_001152763.1 | |||
| NM_001286613.1 | 1520 | Intron | NP_001273542.1 | |||
| NM_022765.3 | 1520 | Intron | NP_073602.3 | |||
| PPIL6 - peptidylprolyl isomerase like 6 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SMPD2 - sphingomyelin phosphodiesterase 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_003080.2 | 1520 | Missense Mutation | ATA,GTA | I,V 376 | NP_003071.2 | |
| XM_005267109.1 | 1520 | Missense Mutation | ATA,GTA | I,V 196 | XP_005267166.1 | |
| XM_011536079.1 | 1520 | Missense Mutation | ATA,GTA | I,V 271 | XP_011534381.1 | |

MICAL1
Genomic Map