Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ASIC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ASIC3 - acid sensing ion channel subunit 3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| CDK5 - cyclin dependent kinase 5 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001164410.2 | 488 | Intron | NP_001157882.1 | |||
| NM_004935.3 | 488 | Intron | NP_004926.1 | |||
| SLC4A2 - solute carrier family 4 member 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001199692.1 | 488 | UTR 5 | NP_001186621.1 | |||
| NM_001199693.1 | 488 | Intron | NP_001186622.1 | |||
| NM_001199694.1 | 488 | Intron | NP_001186623.1 | |||
| NM_003040.3 | 488 | Intron | NP_003031.3 | |||
| XM_006716094.3 | 488 | Intron | XP_006716157.1 | |||

ASIC3
Genomic Map