Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| ABCC6 - ATP binding cassette subfamily C member 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001079528.3 | 2503 | Intron | NP_001072996.1 | |||
| NM_001171.5 | 2503 | Silent Mutation | GCC,GCT | A,A 830 | NP_001162.4 | |
| XM_011522479.2 | 2503 | Silent Mutation | GCC,GCT | A,A 819 | XP_011520781.1 | |
| XM_011522480.1 | 2503 | Silent Mutation | GCC,GCT | A,A 716 | XP_011520782.1 | |
| XM_011522481.2 | 2503 | Silent Mutation | GCC,GCT | A,A 716 | XP_011520783.1 | |
| XM_011522482.2 | 2503 | Silent Mutation | GCC,GCT | A,A 830 | XP_011520784.1 | |
| XM_017023212.1 | 2503 | Silent Mutation | GCC,GCT | A,A 774 | XP_016878701.1 | |
| XM_017023213.1 | 2503 | Silent Mutation | GCC,GCT | A,A 716 | XP_016878702.1 | |
| XM_017023214.1 | 2503 | Silent Mutation | GCC,GCT | A,A 830 | XP_016878703.1 | |
More Information
Set Membership: |
|

ABCC6
Genomic Map