Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC13C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| UNC13C - unc-13 homolog C | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| XM_005254394.4 | Intron | XP_005254451.1 | ||||
| XM_017022220.1 | Intron | XP_016877709.1 | ||||
| XM_017022221.1 | Intron | XP_016877710.1 | ||||
| XM_017022222.1 | Intron | XP_016877711.1 | ||||
| XM_017022223.1 | Intron | XP_016877712.1 | ||||
| XM_017022224.1 | Intron | XP_016877713.1 | ||||
| XM_017022225.1 | Intron | XP_016877714.1 | ||||

UNC13C
Genomic Map