Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS9D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| VPS9D1 - VPS9 domain containing 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004913.2 | 2214 | Silent Mutation | NP_004904.2 | |||
| XM_005256329.4 | 2214 | Missense Mutation | XP_005256386.1 | |||
| XM_005256330.4 | 2214 | Missense Mutation | XP_005256387.1 | |||
| XM_005256331.2 | 2214 | Intron | XP_005256388.1 | |||
| XM_006721350.2 | 2214 | Missense Mutation | XP_006721413.1 | |||
| XM_011523476.2 | 2214 | Missense Mutation | XP_011521778.1 | |||
| XM_011523477.2 | 2214 | Missense Mutation | XP_011521779.1 | |||
| XM_011523478.2 | 2214 | Missense Mutation | XP_011521780.1 | |||
| XM_011523479.2 | 2214 | Missense Mutation | XP_011521781.1 | |||
| XM_011523480.2 | 2214 | Missense Mutation | XP_011521782.1 | |||
| VPS9D1-AS1 - VPS9D1 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| ZNF276 - zinc finger protein 276 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

VPS9D1
Genomic Map