Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| FANCB - Fanconi anemia complementation group B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001018113.2 | Intron | NP_001018123.1 | ||||
| NM_001324162.1 | Intron | NP_001311091.1 | ||||
| NM_152633.3 | Intron | NP_689846.1 | ||||
| XM_011545470.2 | Intron | XP_011543772.1 | ||||
| XM_017029355.1 | Intron | XP_016884844.1 | ||||
| XM_017029356.1 | Intron | XP_016884845.1 | ||||

FANCB
Genomic Map