Assay Details
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||
Literature Links: |
FUS PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
| EUR - Not Available | |||||
| AMR - Not Available |
| FUS - FUS RNA binding protein | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PYCARD - PYD and CARD domain containing | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PYCARD-AS1 - PYCARD antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

FUS
Genomic Map