Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MIR5692C1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MIR5692C1 - microRNA 5692c-1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SLC25A48 - solute carrier family 25 member 48 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_145282.4 | Intron | NP_660325.4 | ||||
| XM_005271903.1 | Intron | XP_005271960.1 | ||||
| XM_006714544.1 | Intron | XP_006714607.1 | ||||
| XM_006714546.1 | Intron | XP_006714609.1 | ||||
| XM_006714550.1 | Intron | XP_006714613.1 | ||||
| XM_006714552.2 | Intron | XP_006714615.1 | ||||
| XM_011543196.1 | Intron | XP_011541498.1 | ||||
| XM_011543199.1 | Intron | XP_011541501.1 | ||||
| XM_017009086.1 | Intron | XP_016864575.1 | ||||
| XM_017009087.1 | Intron | XP_016864576.1 | ||||
| XM_017009088.1 | Intron | XP_016864577.1 | ||||
| XM_017009089.1 | Intron | XP_016864578.1 | ||||
| XM_017009090.1 | Intron | XP_016864579.1 | ||||
| XM_017009091.1 | Intron | XP_016864580.1 | ||||
| XM_017009092.1 | Intron | XP_016864581.1 | ||||

MIR5692C1
Genomic Map