Assay Details
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| GABRA2 - gamma-aminobutyric acid type A receptor alpha2 subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| XM_005248080.3 | Intron | XP_005248137.1 | ||||
| XM_011513675.1 | Intron | XP_011511977.1 | ||||
| XM_011513676.1 | Intron | XP_011511978.1 | ||||
| XM_017007982.1 | Intron | XP_016863471.1 | ||||
| XM_017007983.1 | Intron | XP_016863472.1 | ||||
More Information
Set Membership: |
|

GABRA2
Genomic Map