Assay Details
| CYP2B6 - cytochrome P450 family 2 subfamily B member 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000767.4 | 792 | Missense Mutation | AAG,AGG | K,R 262 | NP_000758.1 | |
More Information
Set Membership: |
|

CYP2B6
Genomic Map