Assay Details
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USH2A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| USH2A - usherin | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_007123.5 | Intron | NP_009054.5 | ||||
| NM_206933.2 | Intron | NP_996816.2 | ||||

USH2A
Genomic Map