Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| NLRP12 - NLR family pyrin domain containing 12 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001277126.1 | Intron | NP_001264055.1 | ||||
| NM_001277129.1 | Intron | NP_001264058.1 | ||||
| NM_144687.3 | Intron | NP_653288.1 | ||||
| XM_011527479.1 | Intron | XP_011525781.1 | ||||
| XM_011527480.1 | Intron | XP_011525782.1 | ||||
| XM_011527482.1 | Intron | XP_011525784.1 | ||||
| XM_017027460.1 | Intron | XP_016882949.1 | ||||
| XM_017027461.1 | Intron | XP_016882950.1 | ||||
| XM_017027462.1 | Intron | XP_016882951.1 | ||||
| XM_017027463.1 | Intron | XP_016882952.1 | ||||
| XM_017027464.1 | Intron | XP_016882953.1 | ||||
| XM_017027465.1 | Intron | XP_016882954.1 | ||||
| XM_017027466.1 | Intron | XP_016882955.1 | ||||
| XM_017027467.1 | Intron | XP_016882956.1 | ||||

NLRP12
Genomic Map