Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| MPP2 - membrane palmitoylated protein 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001278370.1 | Intron | NP_001265299.1 | ||||
| NM_001278371.1 | Intron | NP_001265300.1 | ||||
| NM_001278372.1 | Intron | NP_001265301.1 | ||||
| NM_001278373.1 | Intron | NP_001265302.1 | ||||
| NM_001278374.1 | Intron | NP_001265303.1 | ||||
| NM_001278375.1 | Intron | NP_001265304.1 | ||||
| NM_001278376.2 | Intron | NP_001265305.2 | ||||
| NM_001278381.1 | Intron | NP_001265310.1 | ||||
| NM_005374.4 | Intron | NP_005365.4 | ||||
| XM_011524827.2 | Intron | XP_011523129.2 | ||||
More Information
Set Membership: |
|

MPP2
Genomic Map