Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| PPIL6 - peptidylprolyl isomerase like 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001111298.2 | Intron | NP_001104768.2 | ||||
| NM_001286360.1 | Intron | NP_001273289.1 | ||||
| NM_001286361.1 | Intron | NP_001273290.1 | ||||
| NM_173672.4 | Intron | NP_775943.1 | ||||
| XM_011535765.2 | Intron | XP_011534067.1 | ||||
| XM_011535766.2 | Intron | XP_011534068.1 | ||||
| XM_011535767.2 | Intron | XP_011534069.1 | ||||
| XM_011535769.2 | Intron | XP_011534071.1 | ||||
| XM_017010774.1 | Intron | XP_016866263.1 | ||||
More Information
Set Membership: |
|

PPIL6
Genomic Map