Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF385D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| ZNF385D - zinc finger protein 385D | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_024697.2 | Intron | NP_078973.1 | ||||
| XM_011534122.1 | Intron | XP_011532424.1 | ||||
| XM_011534123.2 | Intron | XP_011532425.1 | ||||
| XM_011534124.2 | Intron | XP_011532426.1 | ||||
| XM_017007191.1 | Intron | XP_016862680.1 | ||||
| XM_017007192.1 | Intron | XP_016862681.1 | ||||
| XM_017007193.1 | Intron | XP_016862682.1 | ||||
| XM_017007194.1 | Intron | XP_016862683.1 | ||||
| XM_017007195.1 | Intron | XP_016862684.1 | ||||
| XM_017007196.1 | Intron | XP_016862685.1 | ||||
| XM_017007197.1 | Intron | XP_016862686.1 | ||||
| XM_017007198.1 | Intron | XP_016862687.1 | ||||
| XM_017007199.1 | Intron | XP_016862688.1 | ||||
| XM_017007200.1 | Intron | XP_016862689.1 | ||||
| XM_017007201.1 | Intron | XP_016862690.1 | ||||
| XM_017007202.1 | Intron | XP_016862691.1 | ||||
| XM_017007203.1 | Intron | XP_016862692.1 | ||||

ZNF385D
Genomic Map