Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LARGE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| LARGE1 - LARGE xylosyl- and glucuronyltransferase 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004737.4 | Intron | NP_004728.1 | ||||
| NM_133642.3 | Intron | NP_598397.1 | ||||
| XM_005261831.3 | Intron | XP_005261888.1 | ||||
| XM_005261832.3 | Intron | XP_005261889.1 | ||||
| XM_011530510.2 | Intron | XP_011528812.1 | ||||
| XM_011530512.2 | Intron | XP_011528814.1 | ||||
| XM_011530513.2 | Intron | XP_011528815.1 | ||||
| XM_011530514.2 | Intron | XP_011528816.1 | ||||
More Information
Set Membership: |
|

LARGE1
Genomic Map