Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986453 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| LOC107986453 - uncharacterized LOC107986453 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| XM_017010130.1 | Intron | XP_016865619.1 | ||||
| XM_017010131.1 | Intron | XP_016865620.1 | ||||
| XM_017010132.1 | Intron | XP_016865621.1 | ||||
| XM_017010133.1 | Intron | XP_016865622.1 | ||||
| XM_017010134.1 | Intron | XP_016865623.1 | ||||
| SLC25A48 - solute carrier family 25 member 48 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_145282.4 | Intron | NP_660325.4 | ||||
| XM_005271903.1 | Intron | XP_005271960.1 | ||||
| XM_006714544.1 | Intron | XP_006714607.1 | ||||
| XM_006714546.1 | Intron | XP_006714609.1 | ||||
| XM_006714550.1 | Intron | XP_006714613.1 | ||||
| XM_006714552.2 | Intron | XP_006714615.1 | ||||
| XM_011543196.1 | Intron | XP_011541498.1 | ||||
| XM_011543199.1 | Intron | XP_011541501.1 | ||||
| XM_017009086.1 | Intron | XP_016864575.1 | ||||
| XM_017009087.1 | Intron | XP_016864576.1 | ||||
| XM_017009088.1 | Intron | XP_016864577.1 | ||||
| XM_017009089.1 | Intron | XP_016864578.1 | ||||
| XM_017009090.1 | Intron | XP_016864579.1 | ||||
| XM_017009091.1 | Intron | XP_016864580.1 | ||||
| XM_017009092.1 | Intron | XP_016864581.1 | ||||
More Information
Set Membership: |
|

LOC107986453
Genomic Map