Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPCML PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| OPCML - opioid binding protein/cell adhesion molecule like | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001012393.2 | Intron | NP_001012393.1 | ||||
| NM_001319103.1 | Intron | NP_001306032.1 | ||||
| NM_001319104.1 | Intron | NP_001306033.1 | ||||
| NM_001319105.1 | Intron | NP_001306034.1 | ||||
| NM_001319106.1 | Intron | NP_001306035.1 | ||||
| NM_002545.4 | Intron | NP_002536.1 | ||||
| XM_006718846.2 | Intron | XP_006718909.1 | ||||
| XM_011542856.2 | Intron | XP_011541158.1 | ||||
More Information
Set Membership: |
|

OPCML
Genomic Map