Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700010L04Rik PubMed Links |
| 1700010L04Rik - RIKEN cDNA 1700010L04 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Gm36584 - predicted gene, 36584 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Mex3b - mex3 homolog B (C. elegans) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_175366.3 | Intron | NP_780575.2 | ||||

1700010L04Rik
Genomic Map