Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2610035D17Rik PubMed Links |
| 2610035D17Rik - RIKEN cDNA 2610035D17 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

2610035D17Rik
Genomic Map