Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700029J07Rik PubMed Links |
| 1700029J07Rik - RIKEN cDNA 1700029J07 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001033148.3 | 1964 | Intron | NP_001028320.1 | |||
| Ccdc110 - coiled-coil domain containing 110 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001033246.2 | 1964 | Silent Mutation | NP_001028418.1 | |||

1700029J07Rik
Genomic Map