Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4430402I18Rik PubMed Links |
| 4430402I18Rik - RIKEN cDNA 4430402I18 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_198651.2 | Intron | NP_941053.2 | ||||
| Slc1a1 - solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_009199.2 | Intron | NP_033225.1 | ||||

4430402I18Rik
Genomic Map