Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
0610040J01Rik PubMed Links |
| 0610040J01Rik - RIKEN cDNA 0610040J01 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_029554.4 | 850 | Missense Mutation | NP_083830.3 | |||
| Rell1 - RELT-like 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_145923.4 | 850 | Intron | NP_666035.2 | |||

0610040J01Rik
Genomic Map