Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700025G04Rik PubMed Links |
| 1700025G04Rik - RIKEN cDNA 1700025G04 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_197990.3 | Intron | NP_932107.1 | ||||

1700025G04Rik
Genomic Map