Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1110004F10Rik PubMed Links |
1110004F10Rik - RIKEN cDNA 1110004F10 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019772.2 | 11 | UTR 5 | NP_062746.1 |
1700003G18Rik - RIKEN cDNA 1700003G18 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
