Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1810041L15Rik PubMed Links |
| 1810041L15Rik - RIKEN cDNA 1810041L15 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001163145.1 | Intron | NP_001156617.1 | ||||

1810041L15Rik
Genomic Map