Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1110046J04Rik PubMed Links |
| 1110046J04Rik - RIKEN cDNA 1110046J04 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Serpinb6a - serine (or cysteine) peptidase inhibitor, clade B, member 6a | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001164117.1 | Intron | NP_001157589.1 | ||||
| NM_001164118.1 | Intron | NP_001157590.1 | ||||
| NM_001243192.1 | Intron | NP_001230121.1 | ||||
| NM_009254.3 | Intron | NP_033280.1 | ||||

1110046J04Rik
Genomic Map