Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2900079G21Rik PubMed Links |
| 2900079G21Rik - RIKEN cDNA 2900079G21 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

2900079G21Rik
Genomic Map