Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1810055G02Rik PubMed Links |
| 1810055G02Rik - RIKEN cDNA 1810055G02 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_028077.2 | 1264 | Silent Mutation | NP_082353.1 | |||
| Gm36608 - predicted gene, 36608 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1810055G02Rik
Genomic Map