Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2610015P09Rik PubMed Links |
| 2610015P09Rik - RIKEN cDNA 2610015P09 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_027801.1 | 1428 | Missense Mutation | NP_082077.1 | |||

2610015P09Rik
Genomic Map