Assay Details
| Species: | Mouse | 
| dbSNP Submissions: | NA | 
| Phenotype: | |
| Literature Links: | 4921507P07Rik PubMed Links | 
| 4921507P07Rik - RIKEN cDNA 4921507P07 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_027564.3 | Intron | NP_081840.1 | ||||
| 5430402O13Rik - RIKEN cDNA 5430402O13 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
 Back To Top
 Back To Top
 4921507P07Rik
4921507P07Rik Genomic Map
Genomic Map Back To Top
 Back To Top