Assay Details
Gene Family ID: | MIPF0000097, mir-338 |
Mature miRNA Sequence: | UCCAGCAUCAGUGAUUUUGUUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Rat | rno-miR-338-3p | MIMAT0000581 | |
| Bovine | bta-miR-338 | MIMAT0009292 | |
| Chicken | gga-miR-338-3p | MIMAT0037552 | |
| Chrysemys picta | cpi-miR-338a-3p | MIMAT0037865 | |
| Columba livia | cli-miR-338b-3p | MIMAT0038621 | |
| Cricetulus griseus | cgr-miR-338 | MIMAT0023918 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-338-3p | MIMAT0047820 | |
| Dog | cfa-miR-338 | MIMAT0006739 | |
| Gorilla gorilla | ggo-miR-338 | MIMAT0024125 | |
| Guinea pig | cpo-miR-338-3p | MIMAT0047175 | |
| Monodelphis domestica | mdo-miR-338 | MIMAT0004136 | |
| Ophiophagus hannah | oha-miR-338-3p | MIMAT0036904 | |
| Pteropus alecto | pal-miR-338-3p | MIMAT0040036 | |
| Python bivittatus | pbv-miR-338-3p | MIMAT0039042 | |
Stem-loop Details
Rat
Bovine
Chicken
Chrysemys picta
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Monodelphis domestica
Ophiophagus hannah
Pteropus alecto
Python bivittatus
More Information
Related Products
miRBase Alias:
Human: hsa-miR-338 (v9.2)
Mouse: mmu-miR-338 (v9.2)
Rat: rno-miR-338 (v18)
Mouse: mmu-miR-338 (v9.2)
Rat: rno-miR-338 (v18)

Genomic Map